ADV412111402 AChemBlock

4-Amino-1-isopropyl-1H-pyrazole

  • MDL number MFCD08700622
  • Cas Number : 97421-16-4
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-1-isopropyl-1h-pyrazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-615
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart