ADV412111404 AChemBlock

4-Bromo-1H-benzoimidazole

  • MDL number MFCD09754164
  • Cas Number : 83741-35-9
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1h-benzoimidazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-617
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart