ADV412111405 AChemBlock

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one

  • MDL number MFCD12755796
  • Cas Number : 943994-02-3
  • Molecular Weight : 275.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-2h-benzo-b-1-4-oxazin-3-4h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-618
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart