ADV412111412 AChemBlock

PHA-848125

  • MDL number MFCD17169990
  • Cas Number : 802539-81-7
  • Molecular Weight : 460.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/pha-848125.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-627
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart