ADV412111428 AChemBlock

Furo[2,3-c]pyridine-5-carboxylic acid

  • MDL number MFCD18255510
  • Cas Number : 478148-62-8
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/furo-2-3-c-pyridine-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-644
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart