ADV412111432 AChemBlock

2,4-Dichloro-5-cyanopyrimidine

  • MDL number MFCD09746248
  • Cas Number : 3177-24-0
  • Molecular Weight : 173.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-5-cyanopyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-647
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart