ADV412111435 AChemBlock

4-Hydroxyisoindolin-1-one

  • MDL number MFCD10000829
  • Cas Number : 366453-21-6
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxyisoindolin-1-one.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-651
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart