ADV412111451 AChemBlock

4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine

  • MDL number MFCD08059805
  • Cas Number : 100644-65-3
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-1h-pyrazolo-3-4-d-pyrimidin-6-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-668
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart