ADV412111454 AChemBlock

6-Bromo-3-iodo-pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD18073604
  • Cas Number : 1109284-33-4
  • Molecular Weight : 323.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-3-iodo-pyrazolo-1-5-a-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-671
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart