ADV412111477 AChemBlock

3-Fluoropropanoic acid

  • MDL number MFCD01695563
  • Cas Number : 461-56-3
  • Molecular Weight : 92.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-fluoropropanoic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-698
pictograms GHS02; GHS07
hstatements H226;H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart