ADV412111482 AChemBlock

4-Amino-6-chloronicotinaldehyde

  • MDL number MFCD09834660
  • Cas Number : 1001756-21-3
  • Molecular Weight : 156.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-amino-6-chloronicotinaldehyde.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-703
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart