ADV412111484 AChemBlock

3-Propylisoxazol-5-amine

  • MDL number MFCD05270911
  • Cas Number : 747411-47-8
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-propylisoxazol-5-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-706
pictograms GHS02; GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart