ADV412111485 AChemBlock

7-Benzyloxy-4-chloro-6-methoxy-quinoline

  • MDL number MFCD12910463
  • Cas Number : 286371-49-1
  • Molecular Weight : 299.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-benzyloxy-4-chloro-6-methoxy-quinoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-707
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart