ADV412111493 AChemBlock

(2R)-1-(Phenylmethyl)-N-[3-(spiro[isobenzofuran-1(3H),4'-piperidin]-1-yl)propyl-2-pyrrolidinecarboxamide

  • MDL number MFCD21607098
  • Cas Number : 475150-69-7
  • Molecular Weight : 433.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-1-phenylmethyl-n-3-spiro-isobenzofuran-1-3h-4-piperidin-1-yl-propyl-2-pyrrolidinecarboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-715
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart