ADV412111495 AChemBlock

6-Chloropyridin-2-ylboronic acid

  • MDL number MFCD08063028
  • Cas Number : 652148-90-8
  • Molecular Weight : 157.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloropyridin-2-ylboronic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-717
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart