ADV412111498 AChemBlock

4-Nitropyridin-2-amine

  • MDL number MFCD04114148
  • Cas Number : 4487-50-7
  • Molecular Weight : 139.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-nitropyridin-2-amine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-722
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart