ADV412111499 AChemBlock

SGX-523

  • MDL number MFCD16660190
  • Cas Number : 1022150-57-7
  • Molecular Weight : 359.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form light pink powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/sgx-523.html

Safety Information

signalword warning
pcodes P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-723
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart