ADV412111513 AChemBlock

2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid

  • MDL number MFCD06659017
  • Cas Number : 5175-14-4
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxo-1-2-dihydro-1-8-naphthyridine-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-737
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart