ADV412111562 AChemBlock

Ethyl morpholine-2-carboxylate hydrochloride

  • MDL number MFCD18911734
  • Cas Number : 135072-31-0
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-morpholine-2-carboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1164
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart