ADV412111572 AChemBlock

8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane

  • MDL number MFCD22035557
  • Cas Number : 1367777-12-5
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-boc-2-oxa-5-8-diazaspiro-3-5-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7426
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart