ADV412111575 AChemBlock

(R)-4-Methyl-2-(hydroxymethyl)morpholine

  • MDL number MFCD16294696
  • Cas Number : 1159598-35-2
  • Molecular Weight : 131.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-4-methyl-2-hydroxymethyl-morpholine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1178
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart