ADV412111589 AChemBlock

(S)-N-Boc-2-(2-oxo-ethyl)-morpholine

  • MDL number MFCD17010134
  • Cas Number : 1257850-93-3
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-n-boc-2-2-oxo-ethyl-morpholine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1192
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart