ADV412111603 AChemBlock

(S)-4-Boc-3-((S)-1-hydroxyethyl)morpholine

  • MDL number MFCD26792479
  • Cas Number : 1821776-37-7
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-4-boc-3-s-1-hydroxyethyl-morpholine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1257
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart