ADV412111611 AChemBlock

(R)-methyl 6,6-dimethyl-morpholine-3-carboxylate

  • MDL number MFCD11053668
  • Cas Number : 1313278-08-8
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form thick liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-methyl-6-6-dimethyl-morpholine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1264
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart