ADV428292809 AChemBlock

(1S,4S)-5-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

  • MDL number MFCD01863510
  • Cas Number : 125224-62-6
  • Molecular Weight : 274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1s-4s-5-methyl-2-5-diazabicyclo-2-2-1-heptane-dihydrobromide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2912
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart