ADV428292817 AChemBlock

exo-7-(Boc-amino)-3-oxa-9-aza-bicyclo[3.3.1]nonane

  • MDL number MFCD28100839
  • Cas Number : 1638683-57-4
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-7-boc-amino-3-oxa-9-aza-bicyclo-3-3-1-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6909
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart