ADV428292823 AChemBlock

5-(3-Pyrrolidinyl)-1H-tetrazole hydrochloride

  • MDL number MFCD28099066
  • Cas Number : 1228583-52-5
  • Molecular Weight : 175.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-3-pyrrolidinyl-1h-tetrazole-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4852-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart