ADV428292827 AChemBlock

exo-3-Oxa-9-aza-bicyclo[3.3.1]nonane-7-acetic acid hemioxalate

  • MDL number MFCD28023900
  • Cas Number : 1523530-64-4
  • Molecular Weight : 460.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/exo-3-oxa-9-aza-bicyclo-3-3-1-nonane-7-acetic-acid-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6860-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart