ADV428292833 AChemBlock

rel-(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrochloride

  • MDL number MFCD26958951
  • Cas Number : 1217827-86-5
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/rel-1s-4s-2-benzyl-2-5-diazabicyclo-2-2-1-heptane-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7762
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart