ADV428292835 AChemBlock

(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-6-methyl-heptanoic acid

  • MDL number MFCD22573387
  • Cas Number : 329270-51-1
  • Molecular Weight : 381.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-methyl-heptanoic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7791
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart