ADV428292840 AChemBlock

4-Chlorothieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD20265442
  • Cas Number : 86825-96-9
  • Molecular Weight : 214.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form brown solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-chlorothieno-2-3-d-pyrimidine-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7860
pictograms GHS07
hstatements H302;H313;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart