ADV428292850 AChemBlock

Methyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD15474972
  • Cas Number : 1062368-70-0
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromopyrazolo-1-5-a-pyridine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2940
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart