ADV428292859 AChemBlock

(2S,4R)-4-aminopyrrolidine-2-carboxylic acid

  • MDL number MFCD03788066
  • Cas Number : 16257-88-8
  • Molecular Weight : 130.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2s-4r-4-aminopyrrolidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-335
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart