ADV428292861 AChemBlock

Azetidine-3-carboxylic acid methyl ester hydrochloride

  • MDL number MFCD01861758
  • Cas Number : 100202-39-9
  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/azetidine-3-carboxylic-acid-methyl-ester-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-420
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart