ADV428292863 AChemBlock

tert-Butyl azetidin-3-ylmethyl(methyl)carbamate hydrochloride

  • MDL number MFCD09998721
  • Cas Number : 1170019-97-2
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-azetidin-3-ylmethyl-methyl-carbamate-hydrochloride.html

Safety Information

signalword Warning
pcodes P260;P264;P270;P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-426
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart