ADV428292867 AChemBlock

6-(p-Toluenesulfonyl)-2-oxa-6-azaspiro[3.3]heptane

  • MDL number MFCD13180735
  • Cas Number : 13573-28-9
  • Molecular Weight : 253.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-p-toluenesulfonyl-2-oxa-6-azaspiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-911
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart