ADV428292870 AChemBlock

tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride

  • MDL number MFCD13185086
  • Cas Number : 1023301-84-9
  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-7-diazaspiro-3-5-nonane-7-carboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1100-1
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart