ADV428292874 AChemBlock

6-Bromoquinoxalin-2(1H)-one

  • MDL number MFCD09834132
  • Cas Number : 55687-34-8
  • Molecular Weight : 225.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromoquinoxalin-2-1h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2701
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart