ADV428292878 AChemBlock

C-Oxetan-2-yl-methylamine

  • MDL number MFCD17214171
  • Cas Number : 882402-12-2
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form colorless liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/c-oxetan-2-yl-methylamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4044
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart