ADV428292889 AChemBlock

(4R,5S)-5-[3,5-Bis(trifluoromethyl)phenyl]-4-methyl-1,3-oxazolidin-2-one

  • MDL number MFCD16294182
  • Cas Number : 875444-10-3
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4r-5s-5-3-5-bis-trifluoromethyl-phenyl-4-methyl-1-3-oxazolidin-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4207
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart