ADV428292895 AChemBlock

Ethyl 6-bromo-1H-pyrazolo[4,3-b]pyridine-3-carboxylate

  • MDL number MFCD17016031
  • Cas Number : 1234616-05-7
  • Molecular Weight : 270.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-bromo-1h-pyrazolo-4-3-b-pyridine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5026
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart