ADV428292898 AChemBlock

(2R)-2-Methyl-4-oxo-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD09832896
  • Cas Number : 790667-43-5
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-2-methyl-4-oxo-piperidine-1-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5134
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart