ADV428292900 AChemBlock

(S)-3-Aminotetrahydrofuran

  • MDL number MFCD08234425
  • Cas Number : 104530-79-2
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-3-aminotetrahydrofuran.html

Safety Information

signalword Danger
pcodes P260;P280;P302+350;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5145
pictograms GHS02; GHS05
hstatements H226;H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart