ADV428292902 AChemBlock

(S)-3-Amino-1-methyl-piperidine dihydrochloride

  • MDL number MFCD20926142
  • Cas Number : 1157849-51-8
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-1-methyl-piperidine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5241
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart