ADV428292915 AChemBlock

(1R,3R,4R)-3-Amino-4-hydroxy-cyclohexanecarboxylic acid ethyl ester hydrochloride

  • MDL number MFCD23703582
  • Cas Number : 1392745-70-8
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-4r-3-amino-4-hydroxy-cyclohexanecarboxylic-acid-ethyl-ester-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5973
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart