ADV428292917 AChemBlock

4,4'-Dibromo-1,1'-binaphthalene

  • MDL number MFCD00021409
  • Cas Number : 49610-35-7
  • Molecular Weight : 412.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-dibromo-1-1-binaphthalene.html

Safety Information

signalword Warning
pcodes P264;P270;P301+312
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5981
pictograms GHS07
hstatements H302;H413
Availability details for
Quantity
  • Availability
  • Add to Cart