ADV428292918 AChemBlock

4-Chlorothieno[3,2-d]pyrimidine-7-carboxylic acid

  • MDL number MFCD20926153
  • Cas Number : 1269667-57-3
  • Molecular Weight : 214.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chlorothieno-3-2-d-pyrimidine-7-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6051
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart