ADV428292921 AChemBlock

6-Bromo-4-chlorothieno[2,3-d]pyrimidine

  • MDL number MFCD09264088
  • Cas Number : 56844-12-3
  • Molecular Weight : 249.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-4-chlorothieno-2-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6158
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart