ADV428292931 AChemBlock

4-Amino-7-bromo-thieno[3,2-d]pyrimidine

  • MDL number MFCD12024855
  • Cas Number : 1318133-32-2
  • Molecular Weight : 230.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-7-bromo-thieno-3-2-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6437
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart