ADV428292939 AChemBlock

Azetidine-3-carboxamide

  • MDL number MFCD10565784
  • Cas Number : 740768-99-4
  • Molecular Weight : 100.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/light-yellow solid/yellowish solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/azetidine-3-carboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2241
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart